MilliporeSigma
All Photos(1)

70001

Sigma-Aldrich

O-[2-(Boc-amino)-ethyl]-O′-[2-(diglycolyl-amino)ethyl]hexaethylene glycol

≥90% (oligomer purity)

Synonym(s):
Boc-PEG-acid (n=7)
Empirical Formula (Hill Notation):
C25H48N2O13
CAS Number:
Molecular Weight:
584.65
MDL number:
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

≥90% (oligomer purity)

form

liquid

reaction suitability

reagent type: cross-linking reagent

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)NCCOCCOCCOCCOCCOCCOCCOCCNC(=O)COCC(O)=O

InChI

1S/C25H48N2O13/c1-25(2,3)40-24(32)26-4-6-33-8-10-35-12-14-37-16-18-39-19-17-38-15-13-36-11-9-34-7-5-27(22(30)20-28)23(31)21-29/h28-29H,4-21H2,1-3H3,(H,26,32)

InChI key

BAMLLPMHTABXAW-UHFFFAOYSA-N

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

SDS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service