702862

Sigma-Aldrich

2-Methoxy-4,4,6-trimethyl-1,3,2-dioxaborinane

Empirical Formula (Hill Notation):
C7H15BO3
CAS Number:
Molecular Weight:
158.00
MDL number:
PubChem Substance ID:

refractive index

n20/D 1.414

density

0.963 g/mL at 25 °C

SMILES string

COB1OC(C)CC(C)(C)O1

InChI

1S/C7H15BO3/c1-6-5-7(2,3)11-8(9-4)10-6/h6H,5H2,1-4H3

InChI key

KASTZDSYCGXZIB-UHFFFAOYSA-N

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

UN 1993C 3 / PGIII

WGK Germany

3

Flash Point(F)

116.6 °F

Flash Point(C)

47 °C

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.