703419

Supelco

Benzamide-15N solution

NMR reference standard, 98 atom % 15N, 2% in DMSO-d6 ("100%", 99.96 atom % D), chromium(III) acetylacetonate 0.2 %, NMR tube size 6.5 mm × 8 in.

Empirical Formula (Hill Notation):
C7H715NO
Molecular Weight:
122.13
MDL number:
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

NMR reference standard
analytical standard

isotopic purity

98 atom % 15N

CofA

certificate is enclosed in each package

composition

chromium(III) acetylacetonate, 0.2%

concentration

2% in DMSO-d6 ("100%", 99.96 atom % D)

application(s)

NMR: suitable

NMR tube size

6.5 mm × 8 in.

format

single component solution

SMILES string

15NH2C(=O)c1ccccc1

InChI

1S/C7H7NO/c8-7(9)6-4-2-1-3-5-6/h1-5H,(H2,8,9)/i8+1

InChI key

KXDAEFPNCMNJSK-VJJZLTLGSA-N

Related Categories

Features and Benefits

Indirect detection test

Quantity

6.5 mm O.D. tube contains 1.2 mL.

RIDADR

UN 1993 / PGIII

WGK Germany

WGK 2

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.