MilliporeSigma
All Photos(1)

704202

Sigma-Aldrich

Nickel oxide - Yttria-stabilized zirconia

greener alternative

for coatings, NiO/YSZ

Linear Formula:
66 wt. % NiO · 34 wt. % (Y2O3ZrO2)

form

powder

composition

NiO, 66 wt. %
(Y2O3-ZrO2), 34 wt. %

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

surface area

spec. surface area 4-8 m2/g (BET)

application(s)

battery manufacturing

Compare Similar Items

View Full Comparison

1 of 4

This Item
544779768359GF65849978
form

powder

form

powder

form

nanopowder

form

powder

composition

NiO, 66 wt. % , (Y2O3-ZrO2), 34 wt. %

composition

(ZrO2)1-x(Y2O3)x

composition

-

composition

-

application(s)

battery manufacturing

application(s)

-

application(s)

-

application(s)

-

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

greener alternative product characteristics

-

surface area

spec. surface area 4-8 m2/g (BET)

surface area

<60 m2/g (BET)

surface area

-

surface area

-

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service