704806

Sigma-Aldrich

1,4-Dioxane solution

NMR reference standard, 1 mM in D2O ("100", 99.96 atom % D), NMR tube size 5 mm × 7 in.

Empirical Formula (Hill Notation):
C4H8O2
CAS Number:
Molecular Weight:
88.11
EC Number:
MDL number:
PubChem Substance ID:
NACRES:
NA.12

Quality Level

grade

NMR reference standard

concentration

1 mM in D2O ("100", 99.96 atom % D)

application(s)

NMR: suitable

NMR tube size

5 mm × 7 in.

SMILES string

C1COCCO1

InChI

1S/C4H8O2/c1-2-6-4-3-5-1/h1-4H2

InChI key

RYHBNJHYFVUHQT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Features and Benefits

13C sensitivity

Quantity

5 mm O.D. tube contains 0.700 mL.

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Certificate of Analysis

Certificate of Origin

Christof Aellig et al.
ChemSusChem, 5(9), 1737-1742 (2012-07-05)
The dehydration of D-fructose to 5-hydroxymethylfurfural was studied under single-phase conditions in the low boiling solvent 1,4-dioxane at moderate temperatures in the presence of the solid acid-catalyst Amberlyst-15. The reaction was first examined and optimized under batch conditions, where it...
Naoyuki Kishimoto et al.
Water research, 47(5), 1919-1927 (2013-02-09)
This paper reports on the reusability and the optimal reuse of iron-rich sludge for an electrochemical Fenton-type process using sequencing batch mode and separation batch mode reuse models. In the sequencing batch mode, processes of electrochemical treatment, neutralization, sedimentation, and...
Sheau-Yun Dora Chiang et al.
Journal of environmental monitoring : JEM, 14(9), 2317-2326 (2012-07-25)
An intrinsic biodegradation study involving the design and implementation of innovative environmental diagnostic tools was conducted to evaluate whether monitored natural attenuation (MNA) could be considered as part of the remedial strategy to treat an aerobic aquifer contaminated with 1,4-dioxane...
Megha Aggarwal et al.
PloS one, 7(12), e51288-e51288 (2012-12-20)
The nucleocapsid core interaction with endodomains of glycoproteins plays a critical role in the alphavirus life cycle that is essential to virus budding. Recent cryo-electron microscopy (cryo-EM) studies provide structural insights into key interactions between capsid protein (CP) and trans-membrane...
A Pan et al.
Langmuir : the ACS journal of surfaces and colloids, 28(39), 13830-13843 (2012-09-07)
Mixed aquo-organic solvents are used in chemical, industrial, and pharmaceutical processes along with amphiphilic materials. Their fundamental studies with reference to bulk and interfacial phenomena are thus considered to be important, but such detailed studies are limited. In this work...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.