704822

Sigma-Aldrich

1,4-Dioxane solution

NMR reference standard, 0.1 mM in D2O ("100%", 99.96 atom % D), NMR tube size 5 mm × 7 in.

Empirical Formula (Hill Notation):
C4H8O2
CAS Number:
Molecular Weight:
88.11
EC Number:
MDL number:
PubChem Substance ID:
NACRES:
NA.12
Pricing and availability is not currently available.

grade

NMR reference standard

concentration

0.1 mM in D2O ("100%", 99.96 atom % D)

application(s)

NMR: suitable

NMR tube size

5 mm × 7 in.

SMILES string

C1COCCO1

InChI

1S/C4H8O2/c1-2-6-4-3-5-1/h1-4H2

InChI key

RYHBNJHYFVUHQT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Features and Benefits

13C sensitivity

Quantity

5 mm O.D. tube contains 0.700 mL.

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin
Venkateshwarlu Gurram et al.
Chemistry, an Asian journal, 7(8), 1853-1861 (2012-05-10)
Reaction conditions for the CC cross-coupling of O(6)-alkyl-2-bromo- and 2-chloroinosine derivatives with aryl-, hetaryl-, and alkylboronic acids were studied. Optimization experiments with silyl-protected 2-bromo-O(6)-methylinosine led to the identification of [PdCl(2)(dcpf)]/K(3)PO(4) in 1,4-dioxane as the best conditions for these reactions (dcpf=1,1'-bis(dicyclohexylphosphino)ferrocene)....
Megha Aggarwal et al.
PloS one, 7(12), e51288-e51288 (2012-12-20)
The nucleocapsid core interaction with endodomains of glycoproteins plays a critical role in the alphavirus life cycle that is essential to virus budding. Recent cryo-electron microscopy (cryo-EM) studies provide structural insights into key interactions between capsid protein (CP) and trans-membrane...
Daniel Gerrity et al.
Water research, 46(19), 6257-6272 (2012-10-16)
The performance of ozonation in wastewater depends on water quality and the ability to form hydroxyl radicals (·OH) to meet disinfection or contaminant transformation objectives. Since there are no on-line methods to assess ozone and ·OH exposure in wastewater, many...
Kyle T Greenway et al.
The Journal of organic chemistry, 77(20), 9221-9226 (2012-10-03)
Hyperconjugation underlies many chemical phenomena of fundamental and practical importance. Owing to a great deal of interest in the anomeric effect, anomeric-like hyperconjugative effects have been thoroughly investigated in oxygen-containing systems. However, such interactions in the second- and third-row chalcogens...
Abdullah M Asiri et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 95, 679-684 (2012-05-18)
The absorption and fluorescence characteristics of 4-(p-N,N-dimethyl-aminophenylmethylene)-2-phenyl-5-oxazolone (DPO) have been investigated in different solvents. DPO dye exhibits a large red shift in both absorption and emission spectra as solvent polarity increases, indicating a large change in the dipole moment of...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.