Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

709441

Sigma-Aldrich

Ubiquitin-15N,D human

98 atom % 15N, 97 atom % D, recombinant, expressed in E. coli

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$3,060.75
10 MG
$5,304.75

$3,060.75

List Price$3,710.00Save 18%
Web-Only Promotion

Check Cart for Availability

Request more information

Select a Size

Change View
5 MG
$3,060.75
10 MG
$5,304.75

About This Item

UNSPSC Code:
12352200
NACRES:
NA.12

$3,060.75

List Price$3,710.00Save 18%
Web-Only Promotion

Check Cart for Availability

Request more information

recombinant

expressed in E. coli

Quality Level

isotopic purity

98 atom % 15N
97 atom % D

form

powder

technique(s)

bio NMR: suitable

storage temp.

−20°C

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
709409709468709395
technique(s)

bio NMR: suitable

technique(s)

bio NMR: suitable

technique(s)

bio NMR: suitable

technique(s)

bio NMR: suitable

isotopic purity

98 atom % 15N, 97 atom % D

isotopic purity

98 atom % 15N

isotopic purity

≥98 atom %

isotopic purity

≥98 atom %

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

powder

form

powder

form

powder

form

powder

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

recombinant

expressed in E. coli

recombinant

expressed in E. coli

recombinant

expressed in E. coli

recombinant

expressed in E. coli

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service