MilliporeSigma
All Photos(1)

711594

Sigma-Aldrich

3-Methyl-2-benzoxazolinone

98%

Synonym(s):
3-Methyl-2(3H)-benzoxazolone
Empirical Formula (Hill Notation):
C8H7NO2
CAS Number:
Molecular Weight:
149.15
Beilstein:
131018
MDL number:
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.5% (GC)
98%

form

needles

mp

83-87 °C

SMILES string

CN1C(=O)Oc2ccccc12

InChI

1S/C8H7NO2/c1-9-6-4-2-3-5-7(6)11-8(9)10/h2-5H,1H3

InChI key

QRMRRLXXFHXMBC-UHFFFAOYSA-N

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service