711896

Sigma-Aldrich

Boc-(R)-3-thienylglycine

98%

Synonym(s):
(R)-2-[(tert-Butoxycarbonyl)amino]-2-(3-thienyl)acetic acid
Empirical Formula (Hill Notation):
C11H15NO4S
CAS Number:
Molecular Weight:
257.31
MDL number:
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

≥97.5% (HPLC)
98%

optical activity

[α]/D -111±5°, c = 1 in ethanol

application(s)

peptide synthesis: suitable

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)N[C@@H](C(O)=O)c1ccsc1

InChI

1S/C11H15NO4S/c1-11(2,3)16-10(15)12-8(9(13)14)7-4-5-17-6-7/h4-6,8H,1-3H3,(H,12,15)(H,13,14)/t8-/m1/s1

InChI key

VJYLMXXKPBZDHN-MRVPVSSYSA-N

Packaging

250 mg in glass bottle
Bottomless glass bottle. Contents are inside inserted fused cone.

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

3

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.