MilliporeSigma
All Photos(1)

712345

Sigma-Aldrich

(2S,2′S)-1,1′-Bis(6-methyl-2-pyridinylmethyl)-2,2′-bipyrrolidine tetrahydrochloride

96%

Synonym(s):
(S,S)-6-Me2-BPBP tetrahydrochloride, 2,2′-[(2S,2′S)-[2,2′-Bipyrrolidine]-1,1′-diylbis(methylene)]bis[6-methylpyridine] tetrahydrochloride
Empirical Formula (Hill Notation):
C22H30N4 · 4HCl
CAS Number:
Molecular Weight:
496.34
MDL number:
PubChem Substance ID:

assay

≥95.5% (HPLC)
95.5-104.5% (calc.on dry substance, AT)
96%

form

lumps

optical activity

[α]/D -10.0±1.0°, c = 1 in H2O

SMILES string

Cl.Cl.Cl.Cl.Cc1cccc(CN2CCC[C@H]2[C@@H]3CCCN3Cc4cccc(C)n4)n1

InChI

1S/C22H30N4.4ClH/c1-17-7-3-9-19(23-17)15-25-13-5-11-21(25)22-12-6-14-26(22)16-20-10-4-8-18(2)24-20;;;;/h3-4,7-10,21-22H,5-6,11-16H2,1-2H3;4*1H/t21-,22-;;;;/m0..../s1

InChI key

ZWSAWWIIHCCUJE-VDUNKYASSA-N

Related Categories

Packaging

500 mg in glass bottle
Bottomless glass bottle. Contents are inside inserted fused cone.

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service