713899

Sigma-Aldrich

(1S,2S)-trans-2-(Boc-amino)-1,2-dihydro-1-naphthol

≥96.0%

Empirical Formula (Hill Notation):
C15H19NO3
CAS Number:
Molecular Weight:
261.32
MDL number:
PubChem Substance ID:

assay

≥96.0% (HPLC)
≥96.0%

optical purity

enantiomeric excess: ≥97.0%

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)N[C@H]1C=Cc2ccccc2[C@@H]1O

InChI

1S/C15H19NO3/c1-15(2,3)19-14(18)16-12-9-8-10-6-4-5-7-11(10)13(12)17/h4-9,12-13,17H,1-3H3,(H,16,18)/t12-,13-/m0/s1

InChI key

KYJBOYBBQNSQCN-STQMWFEESA-N

Related Categories

Packaging

250 mg in glass bottle
Bottomless glass bottle. Contents are inside inserted fused cone.

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service