Skip to Content
MilliporeSigma

718300

5-Bromo-1-methylindole

97%

Synonym(s):

5-Bromo-1-methyl-1H-indole, N-Methyl-5-bromoindole

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C9H8BrN
CAS Number:
Molecular Weight:
210.07
NACRES:
NA.22
PubChem Substance ID:
UNSPSC Code:
12352100
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

5-Bromo-1-methylindole, 97%

InChI

1S/C9H8BrN/c1-11-5-4-7-6-8(10)2-3-9(7)11/h2-6H,1H3

SMILES string

Cn1ccc2cc(Br)ccc12

InChI key

SBOITLSQLQGSLO-UHFFFAOYSA-N

assay

97%

form

solid

mp

39-43 °C

functional group

bromo

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

>230.0 °F - closed cup

flash_point_c

> 110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service