All Photos(1)

720291

Supelco

Ethylbenzene solution

NMR reference standard, 10% in chloroform-d (99.8 atom % D), NMR tube size 3 mm × 8 in.

Empirical Formula (Hill Notation):
C8H10
CAS Number:
Molecular Weight:
106.17
MDL number:
PubChem Substance ID:
NACRES:
NA.12

Quality Level

grade

NMR reference standard

concentration

10% in chloroform-d (99.8 atom % D)

application(s)

NMR: suitable

NMR tube size

3 mm × 8 in.

SMILES string

CCc1ccccc1

InChI

1S/C8H10/c1-2-8-6-4-3-5-7-8/h3-7H,2H2,1H3

InChI key

YNQLUTRBYVCPMQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

1 pkg in ampule

Features and Benefits

1H sensitivity

Quantity

3 mm O.D. tube contains 0.235 mL.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available

Certificate of Analysis

Certificate of Origin

Murthy Kasi et al.
Biodegradation, 24(2), 279-293 (2012-08-16)
A comprehensive study on the effects of different carbon sources during the bacterial enrichment on the removal performances of benzene, toluene, ethylbenzene, and xylenes (BTEX) compounds when present as a mixture was conducted. Batch BTEX removal kinetic experiments were performed...
Dânia Elisa Christofoletti Mazzeo et al.
Chemosphere, 90(3), 1030-1036 (2012-09-18)
The BTEX (benzene, toluene, ethylbenzene and xylene) mixture is an environmental pollutant that has a high potential to contaminate water resources, especially groundwater. The bioremediation process by microorganisms has often been used as a tool for removing BTEX from contaminated...
Carla G A da Silva et al.
Journal of chromatography. A, 1260, 81-87 (2012-09-12)
Endcapped stationary phases were prepared after thermal immobilization of poly(methyloctadecylsiloxane) (PMODS) onto zirconized and titanized silica supports. These new stationary phases have lower densities of residual hydroxyl groups, according to infrared spectroscopy and 29Si CP-MAS NMR and as shown by...
Lili Zhang et al.
Chemosphere, 90(4), 1340-1347 (2012-09-11)
A new strain Mycobacterium cosmeticum byf-4 able to simultaneously degrade benzene, toluene, ethylbenzene, and o-xylene (BTE(o-)X) compounds has been isolated and identified previously in our laboratory. We further report here the extent of degradation of every BTE(o-)X component, and unravel...
Adrián Flores Orozco et al.
Journal of contaminant hydrology, 136-137, 131-144 (2012-07-13)
Broadband spectral induced polarization (SIP) measurements were conducted at a former hydrogenation plant in Zeitz (NE Germany) to investigate the potential of SIP imaging to delineate areas with different BTEX (benzene, toluene, ethylbenzene, and xylene) concentrations. Conductivity images reveal a...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service