Skip to Content
MilliporeSigma

726257

1-Ethyl-3-methylimidazolium hydrogencarbonate solution

~50% in H2O

Synonym(s):

1-Ethyl-3-methylimidazolium bicarbonate solution

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C7H12N2O3
CAS Number:
Molecular Weight:
172.18
UNSPSC Code:
12352100
PubChem Substance ID:
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

InChI

1S/C6H11N2.CH2O3/c1-3-8-5-4-7(2)6-8;2-1(3)4/h4-6H,3H2,1-2H3;(H2,2,3,4)/q+1;/p-1

SMILES string

OC([O-])=O.CCn1cc[n+](C)c1

InChI key

ULMGNXSIBNEBFL-UHFFFAOYSA-M

concentration

~50% in H2O

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

12 - Non Combustible Liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service