Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

726834

Sigma-Aldrich

(S)-Phenyloxirane

ChiPros®, produced by BASF, 98%

Synonym(s):

(S)-Phenylethylene oxide, (S)-Styrene oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8O
CAS Number:
Molecular Weight:
120.15
Beilstein/REAXYS Number:
3587977
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

assay

≥97.5% (GC)
98%

form

liquid

optical purity

enantiomeric excess: ≥97.5%

bp

192-194 °C (lit.)

density

1.051 g/mL at 20 °C (lit.)

SMILES string

C1O[C@H]1c2ccccc2

InChI

1S/C8H8O/c1-2-4-7(5-3-1)8-6-9-8/h1-5,8H,6H2/t8-/m1/s1

InChI key

AWMVMTVKBNGEAK-MRVPVSSYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ChiPros is a registered trademark of BASF SE

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Aquatic Chronic 2 - Carc. 1B - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

174.2 °F - closed cup

flash_point_c

79 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service