732842

Sigma-Aldrich

3-Iodochromone

Synonym(s):
3-Iodo-4H-benzopyran-4-one, 3-Iodo-4H-chromen-4-one
Empirical Formula (Hill Notation):
C9H5IO2
CAS Number:
Molecular Weight:
272.04
MDL number:
PubChem Substance ID:

Quality Level

mp

93-98 °C

SMILES string

IC1=COc2ccccc2C1=O

InChI

1S/C9H5IO2/c10-7-5-12-8-4-2-1-3-6(8)9(7)11/h1-5H

Inchi Key

SZDBDDKQJHHRGH-UHFFFAOYSA-N

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service