733040

Sigma-Aldrich

4-(Aminomethyl)indole

95%

Synonym(s):
NSC 131886
Empirical Formula (Hill Notation):
C9H10N2
CAS Number:
Molecular Weight:
146.19
MDL number:
PubChem Substance ID:

assay

95%

mp

130-135 °C

SMILES string

NCc1cccc2[nH]ccc12

InChI

1S/C9H10N2/c10-6-7-2-1-3-9-8(7)4-5-11-9/h1-5,11H,6,10H2

InChI key

FFBWKPKOXRMLNP-UHFFFAOYSA-N

Application

  • Reactant for preparation of dopamine receptor antagonists
  • Reactant for preparation of high-affinity ligands to α2δ subunit of voltage-gated calcium channels

Packaging

1 g in glass bottle

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

RIDADR

NONH for all modes of transport

WGK Germany

3

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.