733822

Sigma-Aldrich

Trimethyl phosphite solution

NMR reference standard, 1% in acetone-d6 (99.9 atom % D), NMR tube size 3 mm × 8 in.

CAS Number:
MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

grade

NMR reference standard

concentration

1% in acetone-d6 (99.9 atom % D)

application(s)

NMR: suitable

NMR tube size

3 mm × 8 in.

SMILES string

COP(OC)OC

InChI

1S/C3H9O3P/c1-4-7(5-2)6-3/h1-3H3

InChI key

CYTQBVOFDCPGCX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Features and Benefits

31P sensitivity

Quantity

3 mm O.D. tube contains 0.235 mL.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Supp Hazards

EUH066

Hazard Codes

F,Xi

Risk Statement

11-36-66-67

Safety Statement

16-26

RIDADR

UN1090 - class 3 - PG 2 - Acetone, solution

WGK Germany

WGK 3

Flash Point(F)

-2.2 °F

Flash Point(C)

-19 °C

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.