735981

Sigma-Aldrich

Chloroform solution

NMR reference standard, 2% in acetone-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.

MDL number:
PubChem Substance ID:
NACRES:
NA.12
Pricing and availability is not currently available.

grade

NMR reference standard

concentration

2% in acetone-d6 (99.9 atom % D)

application(s)

NMR: suitable

NMR tube size

5 mm × 8 in.

SMILES string

ClC(Cl)Cl

InChI

1S/CHCl3/c2-1(3)4/h1H

InChI key

HEDRZPFGACZZDS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Features and Benefits

1H line shape

Quantity

5 mm O.D. tube contains 0.700 mL.

Signal Word

Danger

Target Organs

Central nervous system

Supp Hazards

EUH066

Hazard Codes

F,Xn

Risk Statement

11-36-40-66-67

Safety Statement

16-26-36/37

RIDADR

UN1090 - class 3 - PG 2 - Acetone, solution

WGK Germany

WGK 3

Flash Point(F)

-2.2 °F - closed cup

Flash Point(C)

-19 °C - closed cup

Certificate of Analysis
Certificate of Origin
Matteo Serino et al.
PloS one, 6(6), e21184-e21184 (2011-06-24)
Daily variations in lipid concentrations in both gut lumen and blood are detected by specific sensors located in the gastrointestinal tract and in specialized central areas. Deregulation of the lipid sensors could be partly involved in the dysfunction of glucose...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.