MilliporeSigma
All Photos(1)

739480

Sigma-Aldrich

2,3-Difluoro-4-(tributylstannyl)pyridine

95%

Empirical Formula (Hill Notation):
C17H29F2NSn
CAS Number:
Molecular Weight:
404.13
MDL number:
PubChem Substance ID:

assay

95%

form

liquid

density

1.233 g/mL at 25 °C

SMILES string

CCCC[Sn](CCCC)(CCCC)c1ccnc(F)c1F

InChI

1S/C5H2F2N.3C4H9.Sn/c6-4-2-1-3-8-5(4)7;3*1-3-4-2;/h1,3H;3*1,3-4H2,2H3;

InChI key

UZBXWEMZNTZREM-UHFFFAOYSA-N

Related Categories

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

SDS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service