748196
solid
[α]/D -138.0°, c = 0.5% in chloroform
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand
105-109 °C
phosphine
20-25°C
C[C@@H](N)[C@H](P(c1ccccc1)c2ccccc2)c3ccccc3
1S/C21H22NP/c1-17(22)21(18-11-5-2-6-12-18)23(19-13-7-3-8-14-19)20-15-9-4-10-16-20/h2-17,21H,22H2,1H3/t17-,21+/m1/s1
JWZAIGGNEGTDMG-UTKZUKDTSA-N
1 of 4
This Item | 716685 | 748242 | 331899 |
---|---|---|---|
form solid | form solid | form solid | form solid |
optical activity [α]/D -138.0°, c = 0.5% in chloroform | optical activity [α]22/D −55.0°, c = 0.5 in chloroform | optical activity [α]22/D 59.0°, c = 0.5% in chloroform | optical activity [α]25/D −7.0°, c = 0.6 in ethanol |
reaction suitability reaction type: Buchwald-Hartwig Cross Coupling Reaction, reaction type: Heck Reaction, reaction type: Hiyama Coupling, reaction type: Negishi Coupling, reaction type: Sonogashira Coupling, reaction type: Stille Coupling, reaction type: Suzuki-Miyaura Coupling, reagent type: ligand | reaction suitability - | reaction suitability - | reaction suitability - |
mp 105-109 °C | mp 120-127 °C | mp 75-81 °C | mp 142-144 °C (lit.) |
functional group phosphine | functional group - | functional group - | functional group - |
storage temp. 20-25°C | storage temp. - | storage temp. - | storage temp. - |
Danger
Skin Corr. 1B
8A - Combustible, corrosive hazardous materials
WGK 3
Not applicable
Not applicable
Enter Lot Number to search for Certificate of Analysis (COA).
Enter Lot Number to search for Certificate of Origin (COO).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service