779814

Sigma-Aldrich

1-Azido-3-fluorobenzene solution

0.5 M in tert-butyl methyl ether, ≥95.0% (HPLC)

Synonym(s):
3-Fluorophenyl azide solution
Empirical Formula (Hill Notation):
C6H4FN3
CAS Number:
Molecular Weight:
137.11
Beilstein/REAXYS Number:
1939302
MDL number:
PubChem Substance ID:

assay

≥95.0% (HPLC)

concentration

0.5 M in tert-butyl methyl ether

impurities

≤2.0% water

storage temp.

−20°C

SMILES string

Fc1cccc(c1)N=[N+]=[N-]

InChI

1S/C6H4FN3/c7-5-2-1-3-6(4-5)9-10-8/h1-4H

InChI key

MQFLRDDKIIPOFC-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Precautionary Statements

RIDADR

UN 2398 3 / PGII

WGK Germany

3

Flash Point(F)

-27.4 °F

Flash Point(C)

-33 °C

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.