MilliporeSigma
All Photos(2)

791040

Sigma-Aldrich

Zinc n-propylsulfinate

95% (H-NMR)

Synonym(s):
Baran NPS Reagent, Bis[(propylsulfinyl)oxy]zinc
Empirical Formula (Hill Notation):
C6H14O4S2Zn
CAS Number:
Molecular Weight:
279.69
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

95% (H-NMR)

form

solid

reaction suitability

reaction type: C-C Bond Formation
reagent type: catalyst
reaction type: C-H Activation

mp

80-105 °C

storage temp.

2-8°C

SMILES string

CCCS(O[Zn]OS(CCC)=O)=O

InChI

1S/2C3H8O2S.Zn/c2*1-2-3-6(4)5;/h2*2-3H2,1H3,(H,4,5);/q;;+2/p-2

InChI key

AVEWLRYVGNMIHD-UHFFFAOYSA-L

Application

Zinc n-propylsulfinate (NPS) is part of a zinc sulfinate toolbox developed by Phil Baran for the simple and rapid diversification of heterocycles.


Learn More at the Professor and Product Portal of Professor Phil S. Baran.

Practical and Innate Carbon-Hydrogen Functionalization of Heterocycles

Packaging

1 g in glass bottle

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service