81559

Sigma-Aldrich

Polystyrene A-NH2

extent of labeling: 0.8-1.2 mmol/g loading

Synonym(s):
(2-Aminoethyl)polystyrene
MDL number:
PubChem Substance ID:

extent of labeling

0.8-1.2 mmol/g loading

matrix

crosslinked with 1% DVB

particle size

500-560 μm

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service