851604

Sigma-Aldrich

Glycyl-L-glutamic acid

Synonym(s):
Gly-Glu
Linear Formula:
HO2CCH2CH2CH(NHCOCH2NH2)CO2H
CAS Number:
Molecular Weight:
204.18
EC Number:
MDL number:

optical activity

[α]22/D −4.7°, c = 5 in H2O

mp

155-158 °C (dec.) (lit.)

SMILES string

NCC(=O)N[C@@H](CCC(O)=O)C(O)=O

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service