Documents

852732

Sigma-Aldrich

6-Methylmercaptopurine

98%

Synonym(s):
6-(Methylthio)purine
Empirical Formula (Hill Notation):
C6H6N4S
CAS Number:
Molecular Weight:
166.20
EC Number:
MDL number:

assay

98%

mp

221-222 °C (lit.)

SMILES string

CSc1ncnc2nc[nH]c12

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service