852872

Sigma-Aldrich

(+)-2′-Deoxyuridine

≥99%

Synonym(s):
1-(2-Deoxy-β-D-ribofuranosyl)uracil, 2′-Deoxyuridine, Uracil deoxyriboside
Empirical Formula (Hill Notation):
C9H12N2O5
CAS Number:
Molecular Weight:
228.20
Beilstein/REAXYS Number:
24433
EC Number:
MDL number:

assay

≥99%

optical activity

[α]26/D +29.8°, c = 2 in H2O

mp

167-169 °C (lit.)

SMILES string

OC[C@H]1O[C@H](C[C@@H]1O)N2C=CC(=O)NC2=O

Application

Used to synthesize enzyme inhibitors, antiviral agents, and anticancer agents.

Certificate of Analysis

Certificate of Origin

Tetrahedron Letters, 35, 1149-1149 (1994)
Aldrichimica Acta, 20, 52-52 (1987)
Lourdes Santana et al.
Journal of medicinal chemistry, 51(21), 6740-6751 (2008-10-07)
The work provides a new model for the prediction of the MAO-A and -B inhibitor activity by the use of combined complex networks and QSAR methodologies. On the basis of the obtained model, we prepared and assayed 33 coumarin derivatives...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service