857289

Sigma-Aldrich

D(+)-Galacturonic acid monohydrate

97%

Empirical Formula (Hill Notation):
C6H10O7 · H2O
CAS Number:
Molecular Weight:
212.15
Beilstein/REAXYS Number:
1727087
MDL number:

assay

97%

SMILES string

O.OC1O[C@@H]([C@H](O)[C@H](O)[C@H]1O)C(O)=O

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service