Sign In to View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C9D6H6
Molecular Weight:
126.23
UNSPSC Code:
12352116
PubChem Substance ID:
Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service
Skip To
isotopic purity
≥98 atom % D
assay
≥98% (CP)
form
liquid
mass shift
M+6
storage temp.
2-8°C
SMILES string
[2H]C([2H])([2H])C1=C(CC([2H])([2H])[2H])C=CC=C1
InChI
1S/C9H12/c1-3-9-7-5-4-6-8(9)2/h4-7H,3H2,1-2H3/i1D3,2D3
InChI key
HYFLWBNQFMXCPA-WFGJKAKNSA-N
Related Categories
1 of 4
This Item | 684198 | 793744 | 709484 |
|---|---|---|---|
| mass shift M+6 | mass shift M+6 | mass shift M+6 | mass shift M+6 |
| isotopic purity ≥98 atom % D | isotopic purity 98 atom % D | isotopic purity 97 atom % D | isotopic purity ≥98 atom % D |
| assay ≥98% (CP) | assay ≥98% (CP) | assay 97% (CP) | assay ≥98% (CP) |
| form liquid | form liquid | form liquid | form liquid |
| storage temp. 2-8°C | storage temp. - | storage temp. 2-8°C | storage temp. - |
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
signalword
Danger
hcodes
Hazard Classifications
Asp. Tox. 1 - Flam. Liq. 3
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
127.4 °F - closed cup
flash_point_c
53 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service




