MilliporeSigma
All Photos(1)

Documents

900981

Sigma-Aldrich

Sodium 4-vinylbenzoic acid

≥98.0%

Sign Into View Organizational & Contract Pricing

Synonym(s):
VBA
Empirical Formula (Hill Notation):
C9H7NaO2
CAS Number:
Molecular Weight:
170.14
MDL number:
NACRES:
NA.23

Assay

≥98.0%

form

liquid

color

colorless to pale yellow

storage temp.

−20°C

Related Categories

General description

Sodium 4-vinylbenzoic acid is a styrenic monomer used to incorporate a pendant carboxylic acid onto a polystyrene backbone. As popular alternatives to acrylic monomers, styrenic monomers generally provide polymers with higher glass transition temperatures, higher modulus, and nominal UV absorbance. Polystyrenes are also generally considered tough and chemically resistant. Styrene monomers are unique as they are one of the few monomers which are able to undergo polymerization by a radical, cationic, or anionic mechanism.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Customers Also Viewed

Slide 1 of 3

1 of 3

Water-Soluble Polymers. 81. Direct Synthesis of Hydrophilic Styrenic-Based Homopolymers and Block Copolymers in Aqueous Solution via RAFT.
Mitsukami Y, et al.
Macromolecules, 34(7), 2248-2256 (2001)
pH-responsive nanofibers with controlled drug release properties.
Demirci S, et al.
Polym. Chem., 5, 2050-2056 (2014)
Polymerization of sodium 4-styrenesulfonate via atom transfer radical polymerization in protic media.
Iddon PD, et al.
Polymer, 45(3), 759-768 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service