901329

Sigma-Aldrich

Bis-MPA-NHBoc dendrimer

trimethylol propane core, generation 3

Synonym(s):
Bis-MPA dendrimer, PFD-G3-TMP-NH-BOC, NHBoc functionalized dendrimer
Empirical Formula (Hill Notation):
C303H494N24O138
Molecular Weight:
6681.24
NACRES:
NA.23

Quality Level

form

solid

mol wt

generation 3

no. Surface Groups

24

color

white to off-white

storage temp.

−20°C

Related Categories

General description

  • Polymer architecture: Dendrimer
  • Generation: 3
  • End Group Functionality: Boc-protected amine
  • No. Surface Groups: 24
  • Calculated Mol. Wt.: 6681.32 g/mol
  • Boc : tert-Butyloxycarbonyl
  • Bis-MPA : 2,2-Bis(hydroxymethyl)propionic acid.

Application

Bis-MPA (or 2,2-Bis(methylol)propionic acid) is an aliphatic, pro-chiral molecule comprised of two hydroxyls and one carboxylic group that has been used in the synthesis of many different types of polymers. Dendrimers synthesized from bis-MPA are biodegradable and have low cytotoxicity, making them well suited for use in biological research applications. These materials have been used in many applications such as signal amplification in bioassays and in drug delivery applications. After Boc deprotection, amine-functionalized dendrimers can be readily used in EDC or DCC coupling reactions with carbonyl-containing compounds to yield highly functionalized materials for a variety of biomedical applications.

storage_class_code

13 - Non Combustible Solids

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Certificate of Origin

Anna Carlmark et al.
Chemical Society reviews, 42(13), 5858-5879 (2013-05-01)
Dendritic polymers are highly branched, globular architectures with multiple representations of functional groups. These nanoscale organic frameworks continue to fascinate researchers worldwide and are today under intensive investigation in application-driven research. A large number of potential application areas have been...
Articles
Professor Michael Malkoch provides an overview of the unique properties of dendritic molecules and their application in biomedical fields. He highlights bis-MPA dendritic scaffolds as a promising biodegradable and biocompatible platform for drug delivery applications
Read More

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service