901553
powder
white to pale yellow
conforms to structure for NMR
2-8°C
1 of 4
This Item | 900496 | 900622 | 900629 |
---|---|---|---|
form powder | form powder | form powder | form powder |
color white to pale yellow | color - | color - | color - |
suitability conforms to structure for NMR | suitability - | suitability - | suitability - |
storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C |
13 - Non Combustible Solids
WGK 3
Not applicable
Not applicable
Enter Lot Number to search for Certificate of Analysis (COA).
Enter Lot Number to search for Certificate of Origin (COO).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service