Skip to Content
MilliporeSigma

Skip To

904538

DMT-2′O-TBDMS-rA(ac)-1-15N phosphoramidite

≥98 atom % 15N, ≥95% (CP)

Synonym(s):

N-acetyl-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-O-[(1,1-dimethylethyl)dimethylsilyl]-adenosine-1-15N, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite]

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 mg
Check Cart for Availability
$651.00

About This Item

Empirical Formula (Hill Notation):
C48H6415NN6O8PSi
Molecular Weight:
927.12
MDL number:
UNSPSC Code:
41106305
NACRES:
NA.12
Isotopic purity:
≥98 atom % 15N
Assay:
≥95% (CP)
Mass shift:
M+1
Form:
powder

$651.00


Check Cart for Availability

Request more information
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


isotopic purity

≥98 atom % 15N

Quality Segment

assay

≥95% (CP)

form

powder

technique(s)

bio NMR: suitable, oligo synthesis: suitable

mass shift

M+1

storage temp.

−20°C

SMILES string

CC(NC1=[15N]C=NC2=C1N=CN2[C@H]3[C@H](O[Si](C(C)(C)C)(C)C)[C@H](OP(N(C(C)C)C(C)C)OCCC#N)[C@@H](COC(C4=CC=CC=C4)(C5=CC=C(OC)C=C5)C6=CC=C(OC)C=C6)O3)=O

InChI key

IISGHHSYMWMWKH-HLCWUODISA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
904554904635904562
assay

≥95% (CP)

assay

≥95% (CP)

assay

≥95% (CP)

assay

≥95% (CP)

technique(s)

bio NMR: suitable, oligo synthesis: suitable

technique(s)

bio NMR: suitable, oligo synthesis: suitable

technique(s)

bio NMR: suitable, oligo synthesis: suitable

technique(s)

bio NMR: suitable, oligo synthesis: suitable

isotopic purity

≥98 atom % 15N

isotopic purity

≥98 atom % 15N

isotopic purity

≥98 atom % 15N

isotopic purity

≥98 atom % 15N

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

powder

form

powder

form

powder

form

powder

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


flash_point_c

Not applicable

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters