909211

Sigma-Aldrich

Cerium doped iron oxide

Synonym(s):
Ce3/4+ doped iron oxide nanoparticles, Ce3/4+ doped maghemite nanoparticles

concentration

3.2 mg/mL in H2O

particle size

6-10 nm

Looking for similar products? Visit Product Comparison Guide

Application

Functional shell formation with ceric ammonium nitrate (CAN, Ce-(NH4)2(NO3)6) on nanometer sized maghemite (γ-Fe2O3) particles has been shown to promote the binding of oxidized polyethyleneimine (PEI) to their surface, which mitigates their acute in vivo toxicity and enables the applications of cerium doped maghemite nanoparticles in high efficacy gene silencing. In addition these nanoparticles find applications in magnetic resonance imaging (MRI) and drug delivery.

Legal Information

The Product is being introduced through Materials Transfer Agreement with ′Bar Ilan University, Israel′.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

WGK Germany

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin
Ce3/4+ cation-functionalized maghemite nanoparticles towards siRNA-mediated gene silencing
Israel L L et al.
Journal of Material Chemistry B: Materials for Biology and Medicine, 2, 6215-6225 (2014)
Liron Limor Israel et al.
ACS applied materials & interfaces, 7(28), 15240-15255 (2015-06-30)
A ceric ammonium nitrate (CAN)-based doping step was used for the fabrication of core maghemite nanoparticles (NPs) that enabled the obtainment of colloid particles with a view to a high-level nanoparticle (NP) surface doping by Ce(III/IV). Such doping of Ce(III/IV)...
E Lellouche et al.
Bioconjugate chemistry, 26(8), 1692-1701 (2015-06-10)
Silencing of RNA to knock down genes is currently one of the top priorities in gene therapies for cancer. However, to become practical the obstacle of RNA delivery needs to be solved. In this study, we used innovative maghemite (γ-Fe2O3)...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.