Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

916668

Sigma-Aldrich

PLAM-AβIδ1MεLVproS-13CH3 Methyl Labeling Kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$1,460.00

$1,460.00


Estimated to ship onMay 14, 2025


Request more information

Select a Size

Change View
1 KIT
$1,460.00

About This Item

UNSPSC Code:
12352200

$1,460.00


Estimated to ship onMay 14, 2025


Request more information

technique(s)

bio NMR: suitable

Quality Level

Related Categories

General description

PLAM-AβIδ1MεLVproS-13CH3 kit has 13CH3 isotopomer precursors and contains protocol instructions for creation of isotopically-labeled proteins.

Application

For detection of long-range nOes or for the NMR study of large proteins
PLAM-AβIδ1MεLVproS-13CH3 is an optimized kit to produce proteins with 13CH3 isotopomer on the methyl groups of alanine, isoleucine, methionine, leucine, and valine. This kit has been tested with protein isotopic labeling in E. coli. It is used in NMR investigations of the structure, dynamics and function of challenging biological systems at atomic resolution. Introduction of 13C -labeled methyl group in highly deuterated background enhances the resolution of methyl-TROSY spectra.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service