MilliporeSigma
All Photos(1)

91679

Sigma-Aldrich

N-(6-Hydroxyhexyl)trifluoroacetamide

≥98.0% (GC)

Synonym(s):
6-(Trifluoroacetamido)-1-hexanol
Empirical Formula (Hill Notation):
C8H14F3NO2
CAS Number:
Molecular Weight:
213.20
Beilstein:
1949232
MDL number:
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

≥98.0% (GC)

form

crystals

reaction suitability

reagent type: cross-linking reagent

mp

46-49 °C

SMILES string

OCCCCCCNC(=O)C(F)(F)F

InChI

1S/C8H14F3NO2/c9-8(10,11)7(14)12-5-3-1-2-4-6-13/h13H,1-6H2,(H,12,14)

InChI key

BGCYSPBEPMOQII-UHFFFAOYSA-N

Packaging

1 g in glass bottle

Other Notes

Employed for preparing libraries of oligosaccharides with an amino tether

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

SDS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service