Skip to Content

916919

QLAM-Iδ1MεLVproS-13CH3 Methyl Labeling Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352200
Technique(s):
bio NMR: suitable
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


technique(s)

bio NMR: suitable

Quality Segment

General description

QLAM-Iδ1MεLVproS-13CH3 kit has 13CH3 isotopomer precursors and contains protocol instructions for creation of isotopically-labeled proteins.

Application

For detection of long-range nOes or for the NMR study of large proteins
QLAM-Iδ1MεLVproS-13CH3 is an optimized kit to produce proteins with 13CH3 isotopomer on the methyl groups of isoleucine, methionine, leucine, and valine. This kit has been tested with protein isotopic labeling in E. coli. It is used in NMR investigations of the structure, dynamics and function of challenging biological systems at atomic resolution. Introduction of 13C -labeled methyl group in highly deuterated background enhances the resolution of methyl-TROSY spectra.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact
Stable Isotopes Customer Service.



pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library