Skip to Content
MilliporeSigma

925373

Sigma-Aldrich

3-epi-25-Hydroxyvitamin D2-(22,26,27-13C3) solution

10 μg/mL in ethanol, ≥98 atom % 13C, ≥95% (CP)

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$733.00

$733.00


Check Cart for Availability

Request more information

About This Item

Empirical Formula (Hill Notation):
13C3C25H44O2
Molecular Weight:
415.63
UNSPSC Code:
12352005
NACRES:
NA.13

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

isotopic purity

≥98 atom % 13C

Quality Level

assay

≥95% (CP)

concentration

10 μg/mL in ethanol

mass shift

M+3

SMILES string

C[C@@]12[C@](CC[C@]2([H])[C@H](C)/[13CH]=C/[C@H](C)C(O)([13CH3])[13CH3])([H])/C(CCC1)=C/C=C(C[C@H](O)CC3)\C3=C

InChI

1S/C28H44O2/c1-19-10-14-24(29)18-23(19)13-12-22-8-7-17-28(6)25(15-16-26(22)28)20(2)9-11-21(3)27(4,5)30/h9,11-13,20-21,24-26,29-30H,1,7-8,10,14-18H2,2-6H3/b11-9+,22-12+,23-13-/t20-,21+,24-,25-,26+,28-/m1/s1/i4+1,5+1,9+1

InChI key

KJKIIUAXZGLUND-BOKKBJPHSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
924881925357924865
isotopic purity

≥98 atom % 13C

isotopic purity

≥98 atom % 13C

isotopic purity

≥98 atom % 13C

isotopic purity

≥98 atom % 13C

assay

≥95% (CP)

assay

≥95% (CP)

assay

≥95% (CP)

assay

≥95% (CP)

mass shift

M+3

mass shift

M+3

mass shift

M+5

mass shift

M+5

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

concentration

10 μg/mL in ethanol

concentration

10 μg/mL in ethanol

concentration

10 μg/mL in ethanol

concentration

10 μg/mL in ethanol

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

pictograms

FlameExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

55.4 °F - closed cup

flash_point_c

13 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service