Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

931675

Sigma-Aldrich

Fmoc-L-H3Pyrd6OMe-OH

Novabiochem®, ≥95%

Synonym(s):

(αS)-α-[[(9H-Fluoren-9-ylmethoxy)carbonyl]amino]-6-methoxy-3-pyridazinebutanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H23N3O5
CAS Number:
Molecular Weight:
433.46
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

assay

≥95%

form

powder

mp

116-126 °C

storage temp.

−20°C

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
931659931616931780
assay

≥95%

assay

≥95%

assay

≥95%

assay

≥95%

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

mp

116-126 °C

mp

-

mp

193-195 °C

mp

233-235 °C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

form

powder

form

crystals or chunks

form

powder

form

powder

Application

Fmoc-L-H3Pyrd6OMe-OH is an arginine analog that can be used to prepare novel peptides for usage as putative therapeutics or in the elucidation of structure-activity relationships.

Legal Information

Product of Peptidream
Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service