Skip to Content
MilliporeSigma

931764

Fmoc-L-K4COpip1aa(tBu)-OH

Novabiochem®, ≥95 %

Sign In to View Organizational & Contract Pricing.

Select a Size

Pricing and availability is not currently available.

About This Item

Empirical Formula (Hill Notation):
C33H43N3O7
Molecular Weight:
593.71
NACRES:
NA.21
UNSPSC Code:
12352209
Assay:
≥95 %
Form:
powder

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Fmoc-L-K4COpip1aa(tBu)-OH, Novabiochem®, ≥95 %

assay

≥95 %

form

powder

storage temp.

−20°C

Quality Level

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
931659931616931780
assay

≥95 %

assay

≥95%

assay

≥95%

assay

≥95%

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

form

powder

form

crystals or chunks

form

powder

form

powder

Application

Fmoc-L-K4COpip1aa(tBu)-OH is an arginine analog that can be used to prepare novel peptides for usage as putative therapeutics or in the elucidation of structure-activity relationships.

Legal Information

Product of Peptidream
Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service