MilliporeSigma
All Photos(1)

94042

Sigma-Aldrich

Dimethyl undecanedioate

technical, ≥90% (GC)

Linear Formula:
CH3OCO(CH2)9COOCH3
CAS Number:
Molecular Weight:
244.33
Beilstein:
1789014
EC Number:
MDL number:
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Quality Level

assay

≥90% (GC)

form

liquid

refractive index

n20/D 1.440

bp

123-124 °C/2 mmHg (lit.)

mp

17-19 °C

density

0.983 g/mL at 20 °C (lit.)

SMILES string

COC(=O)CCCCCCCCCC(=O)OC

InChI

1S/C13H24O4/c1-16-12(14)10-8-6-4-3-5-7-9-11-13(15)17-2/h3-11H2,1-2H3

InChI key

LADJFIHHYMBJHB-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service