Skip to Content
MilliporeSigma

940631

Nickel(II) acetate

(anhydrous), ≥99.9% trace metals basis (basis)

Synonym(s):

Nickel acetate, Nickel(2+) acetate, Nickelous acetate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C4H6NiO4
CAS Number:
Molecular Weight:
176.78
NACRES:
NA.21
UNSPSC Code:
11101706
Assay:
(EDTA titration), ≥99.9% trace metals basis (basis)
Form:
powder, crystals or chunks
Solubility:
water: soluble
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

(EDTA titration), ≥99.9% trace metals basis (basis)

form

powder, crystals or chunks

solubility

water: soluble

anion traces

chloride (Cl-): ≤50 ppm, sulfate (SO42-): ≤50 ppm

cation traces

Al: ≤10 ppm, Ca: ≤10 ppm, Cd: ≤10 ppm, Cr: ≤10 ppm, Cu: ≤10 ppm, Fe: ≤10 ppm, K: ≤10 ppm, Mg: ≤10 ppm, Na: ≤30 ppm, Pb: ≤10 ppm, Si: ≤10 ppm, Zn: ≤10 ppm

SMILES string

CC(=O)[O-].CC(=O)[O-].[Ni+2]

InChI

InChI=1S/C2H4O2.Ni/c1-2(3)4;/h1H3,(H,3,4);

InChI key

XMOKRCSXICGIDD-UHFFFAOYSA-N

General description

Nickel(II) Acetate, Anhydrous is a water-soluble crystalline compound with solubility in methanol. We provide it for research purposes with a purity of ≥ 99.9%, anhydrous, and battery grade. It features low sulfate and chloride content and lower levels of metal traces. Our Nickel(II) Acetate, Anhydrous, ≥99.9% trace metals basis serves as a suitable precursor for the synthesis of Cathode active materials through the sol-gel synthesis method.

Application

Nickel(II) Acetate, anhydrous, finds extensive use in research and development (R&D), particularly in the field of chemistry for the synthesis of nanomaterials, composites, electrodes, and catalysts across various applications. Specifically, Nickel(II) Acetate, anhydrous, battery grade is the perfect choice for battery applications in synthesizing cathodes using various synthesis routes.


For example: Nickel(II) acetate, anhydrous, is employed in the synthesis of spinel Lithium Nickel Manganese Oxide (LNMO) using a precipitation method, in the presence of Manganese(II) Acetate and Manganese Oxalate in a microemulsion system. The resulting porous nanorods of spinel-type LiNi0.5Mn1.5O4, composed of assembled nanoparticles, have been investigated as cathode materials for rechargeable lithium-ion batteries, demonstrating excellent performance in high-rate capability and long cycle life. In one of the studies, it shows that Nickel(II) Acetate, anhydrous, in conjunction with tetrabutyl titanate, has been utilized as a precursor in the presence of ethylene glycol to synthesize porous NiTiO3 nanorods using an ethylene glycol-mediated route, followed by calcination at 600 °C at room temperature. These porous nanorods have demonstrated excellent performance in the visible light photocatalytic degradation of nitrobenzene, making them a promising material for visible light photocatalysis applications. Furthermore, Nickel(II) Acetate, anhydrous, has been studied for its application as a precursor in the synthesis of N-doped sponge nickel, comprising interconnected Ni micro/nanofibers, prepared through a hydrothermal method using nickel acetate followed by annealing in NH3. This unique catalyst exhibits a 3D porous structure and high electronic conductivity, making it suitable for the oxygen evolution reaction (OER) in applications such as water splitting and rechargeable metal-air batteries.. Nickel(II) acetate anhydrous Interestingly, Nickel(II) Acetate, anhydrous, has found application in polymer solar cells utilizing poly(3-hexylthiophene) (P3HT) and indene-C60 bisadduct (ICBA), where it is employed as the hole collection layer. After thermal annealing, nickel acetate demonstrates high transparency, suitable energy levels, and a high hole mobility, establishing its potential as an alternative to conventional PEDOT:PSS for the hole collection layer in these solar cells.

Features and Benefits

1.Anhydrous form with low moisture level (< 0.5%)
2.Suitable for battery and solar cell applications due to low moisture content
3.Low anion content (chloride & sulfate)
4.High purity of 99.9% (<1000 ppm)
5. Similar Specification like battery grade


Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library