Skip to Content
MilliporeSigma

943886

Conductive Silver Nanoink for Inkjet Printing

25%, bio-based

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

Surfactant: Oleylamine
Solvent, solvent mixtures Hydrocarbon, proprietary

Quality Segment

form

liquid

concentration

25%

particle size

8-20 nm

storage temp.

2-30°C

Application

Introducing our advanced silver nanoink, specifically engineered for inkjet printing across a variety of substrates. This high-performance ink features silver nanoparticles with an average size of around 10 nm, ensuring superior conductivity and adhesion. With a silver concentration of approximately 25 wt/vol% and a viscosity of about 11 cP, its unique formulation allows for excellent solubility in nonpolar organic solvents, providing stability under atmospheric conditions at room temperature. Ideal for use on materials such as paper, silicon, and polycarbonate, our silver nanoink is particularly suitable for applications requiring fine line printing due to its low sintering temperature and outstanding resistivity results. Perfect for both industrial and creative applications, this cutting-edge nanoink technology is designed to enhance your projects with precision and reliability.

Disclaimer

This product is not intended for antimicrobial purpose.


signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2

target_organs

Nervous system

Storage Class

3 - Flammable liquids

flash_point_f

125.6 °F

flash_point_c

52 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library