Skip to Content
MilliporeSigma

943916

Conductive Gold Nanoink for Aerosol-Jet printing

Bio-based

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

description

Printable Material: ITO, Glass, Kapton, Resistivity: 2 -7 x of bulk gold depending on T/time, Solvent: Non-polar solvent formulations, Surfactant: Oleylamine

form

liquid

concentration

62-72%

particle size

~3-5 nm

viscosity

~7-10 cP

storage temp.

2-30°C

Application

Our gold nanoink is a general-use ink for Ink-Jet printing on variety of substrates. The ink is based on gold nanoparticles with average size around 4 nm and dispersed in liquid vehicle. Since it is surface stabilized, ink is highly soluble in nonpolar organic solvent and stable under atmospheric conditions at room temperature.

Preparation Note

General recommendation: Stored under inert atmosphere curing temperature: >~200 - 400oC

Other Notes

FAQs: Q1: What adjustments can improve sintering results, especially with older ink? Answer: Try increasing the sintering duration or slightly raising the temperature. These adjustments can enhance results when using older ink.


Q2: How does storage duration affect the ink’s condition? Answer: If the ink has been stored for around seven months, its condition can vary depending on how it was stored.


Q3: Is ink still usable after being refrigerated for several months? Answer: Yes, refrigerated ink is generally still usable. However, it may require longer or higher-temperature sintering to achieve optimal performance.


Q4: What can be done if the ink contains bulk material? Answer: You can centrifuge the ink at a low speed (around 500 rpm) to help remove any bulk material.


Q5: What degradation risks are there if the ink was not sealed or sparged?

Answer: Without proper sealing or sparging (using Argon or Nitrogen), the ink may degrade over time. This could result in reduced concentration. Centrifuging can help, but performance may still be affected.


Q6: What is the recommended storage method for ink kept longer?

Answer: Sparge the ink with Argon or Nitrogen and seal it with electrical tape to preserve quality during extended storage. For best results, sparging the bottle with argon to displace oxygen and moisture is recommended, along with ensuring a secure seal. Periodically inverting the bottle to remix the contents helps maintain uniformity and prevents settling.


signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1

Storage Class

3 - Flammable liquids

flash_point_f

125.6 °F

flash_point_c

52 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library