Skip to Content
MilliporeSigma

945730

Tyramine functionalized gelatin

40% functionalized

Synonym(s):

Fish gelatin, Gelatin tyramine

Sign In to View Organizational & Contract Pricing.

Select a Size

Pricing and availability is not currently available.

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

SMILES string

[*]NC(C)C(NCC(N1CCCC1C(NC(CCCCNC(CCC2=CC=C(O)C=C2)=O)C(NCC(NC(CCC([O-])=O)C(N3C(C(N[*])=O)CC(O)C3)=O)=O)=O)=O)=O)=O

biological source

fish

form

powder or chunks (or fibers)

Quality Level

color

white to faint yellow-brown

application(s)

3D bioprinting
tissue engineering
(Hydrogels)

storage temp.

−20°C

General description

Tyramine functionalized gelatin has garnered significant attention in biomedical applications due to its unique properties that enhance biocompatibility and bioactivity. Gelatin, a natural polymer derived from collagen, possesses excellent biocompatibility, biodegradability, and low immunogenicity, making it a suitable scaffold material for tissue engineering. The introduction of tyramine groups into gelatin enhances its functionality by allowing for improved cross-linking and interaction with biological molecules. This modification facilitates the development of hydrogels that can mimic the extracellular matrix, providing a supportive environment for cell adhesion, proliferation, and differentiation.

Application

The versatility of the tyramine functionalized gelatin positions it as a promising material for advancing biomedical technologies including:
  • Drug delivery systems - Tyramine functionalized gelatin can be utilized to create responsive carriers that release therapeutic agents in a controlled manner. The presence of tyramine enables the formation of covalent bonds with various drugs, enhancing their stability and release profiles.
  • Regenerative medicine - Tyramine modified gelatin can be tailored to respond to specific stimuli such as pH or temperature changes, allowing for targeted delivery in various biomedical applications including regenerative medicine and tissue engineering.

Features and Benefits

  • Biocompatible and biodegradable
  • Tyramine functionalized for improved crosslinking
  • Mimics native extracellular environment to improve and support cell-matrix interactions

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service