Skip to Content
MilliporeSigma

945773

Cellulose Nanocrystals

5 wt. % solids in water

Synonym(s):

CNC, Nanocellulose

Sign In to View Organizational & Contract Pricing.

Select a Size

Pricing and availability is not currently available.

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

InChI

InChI=1S/H2O/h1H2

SMILES string

O[C@H]1[C@H](O[C@@H]2[C@@H](CO)O[C@@H](O[H])[C@H](O)[C@H]2O)O[C@H](CO)C(O)[C@@H]1O

form

liquid

concentration

5 wt. % solids in water

color

colorless to brown

application(s)

advanced drug delivery
(Biocomposites)

storage temp.

2-8°C

Quality Level

General description

Cellulose is considered as one of the most ubiquitous structural renewable polymeric resources. Cellulose nanocrystals (CNCs) are the nanometric dimensions of celluloses, typically measuring a few nanometers to 50 nanometers in diameter and reaching several micrometers in length. We offer the product with diamter ~ 10 nm to 50 nm and length ~ 120 nm.

Application

The product can be used for biocomposites[[1]], drug delivery, coating and packaging applications. By introducing a diverse array of functional groups, CNCs can selectively bind to oppositely charged drugs or macromolecules, enhancing their capability for effective carrying, delivery, and controlled release. Furthermore, this chemical modification not only boosts the biocompatibility of Cellulose nanocrystals (CNCs) but also improves their stability in biological environments. The ability to tailor the surface charge and chemistry of CNCs positions them as versatile substrates, ideal for advancing biomedical and drug delivery applications. [[2],[3]]

Features and Benefits

  • High mechanical strength, better reinforcing capabilities
  • Easy to functionalize for various applications
  • High degree of crystallinity
  • Low density with large surface area

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service