Skip to Content
MilliporeSigma

945781

Methoxy-poly(ethylene glycol) amine-block-poly-L-lysine hydrobromide

Mn 5000 (mPEG), Mn 15000 (PLL)

Synonym(s):

mPEG-amine5k-PLL15k, mPEGNH2-PLL

Sign In to View Organizational & Contract Pricing.

Select a Size

Pricing and availability is not currently available.

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

SMILES string

COCCOCCNC(C(CCCC[NH3+])N[H])=O.[Br-]

form

powder or chunks

mol wt

Mn 15000 (PLL), Mn 5000 (mPEG)

color

white

application(s)

(Drug Delivery)

storage temp.

−20°C

Quality Level

General description

Methoxy poly(ethylene glycol) amine-block-poly(L-lysine) (mPEG-PLL) is a cationic diblock copolymer consiting of amine-functionalized PEG and PLL that can be used in drug delivery applications. The cationic PLL block can bind to DNA or RNA, as well as other therapuetic agents, while the amine-functionalized PEG block can be further functionalized with targeting moieties for targeted and controlled drug delivery. This cationic diblock copolymer can self-assemble in the presence of a drug to form polymeric micelles and polymersomes.

Application

  • Non-viral gene delivery application
  • Targeted drug delivery application
  • Protein coatings and tissue culture

Features and Benefits

  • Self-assemble capability to form micelles and polymersomes
  • Amine functional group at the side chain enables further rapid functionalization
  • Cationic poly(L-lysine) has excellent DNA and RNA condensation property
  • Biodegradable, biocompatible and water soluble

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service