Skip to Content
MilliporeSigma

945811

Poly(butynyl phospholane)

Mn 10,000

Synonym(s):

PByP

Sign In to View Organizational & Contract Pricing.

Select a Size

Pricing and availability is not currently available.

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

SMILES string

COCCOP(OCCC#C)(C)=O

form

liquid

mol wt

Mn 10,000

color

yellow to orange

application(s)

advanced drug delivery

storage temp.

2-8°C

Quality Level

General description

Poly(phosphoester)s (PPEs) are interesting candidates as biomimetic building blocks for biocompatible and biodegradable polymers, as their backbone is recognizable to enzymes and can be cleaved or built up under physiological conditions.

Application

  • The development of nanocariers for drug and gene delivery application
  • Fabrication of hydrogels for tissue engineering
  • In the drug delivery system

Features and Benefits

  • Biocompatible and biodegradable polymers can be synthesized by ROP
  • Exhibits thermoresponsive behavior (LCST) in aqueous solution
  • Degradable polymers at different pH level
  • Hydrophobic drug conjucation site is available in the side chain

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service