Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now

945854

Methoxy-poly(ethylene glycol)-block-poly(L-aspartic acid sodium salt)

mPEG 5k, poly-L-aspartic acid sodium salt 15k

Synonym(s):

mPEG-PASP

Sign In to View Organizational & Contract Pricing.

Select a Size


Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

mol wt

256,23 g/mol

color

white to light yellow-brown

application(s)

(Drug delivery)

storage temp.

−20°C

SMILES string

COCCOCCNC(C(CC(O[Na])=O)N[H])=O

form

powder, flakes, or chunks

Quality Level

General description

Poly(L-aspartic acid sodium salt) is an anionic polymer which has protein like backbone structure. Due to its high biodegradability, poly(L-aspartic acid sodium salt) has been copled with different synthetic degradable polymer like methoxy PEG or PLA. Those copolymers can form micellar nanostructure which can be used for drug delivery application.

Application

  • Drug delivery application
  • DNA delivery
  • Bone targeting application
  • Surface modification

Features and Benefits

  • Biocompatible and biodegradable
  • High calcium affinity or Calcium chelation ability

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service