Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now

945943

N-(3,4-Dihydroxyphenethyl)methacrylamide

≥95%

Synonym(s):

N-(3,4-dihydroxyphenethyl)methacrylamide, N-[2-(3,4-Dihydroxyphenyl)ethyl]-2-methyl-2-propenamide, N-[2-(3,4-Dihydroxyphenyl)ethyl]-2-methylacrylamide, DMA, Dopamine Methacrylamide, Dopamine methacrylate

Sign In to View Organizational & Contract Pricing.

Select a Size


Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

InChI

1S/C12H15NO3/c1-8(2)12(16)13-6-5-9-3-4-10(14)11(15)7-9/h3-4,7,14-15H,1,5-6H2,2H3,(H,13,16)

SMILES string

O=C(NCCC1=CC=C(O)C(O)=C1)C(=C)C

assay

≥95%

form

solid

color

white to yellow

storage temp.

2-8°C

Quality Level

General description

N-(3,4-Dihydroxyphenethyl)methacrylamide (DMA) is a specialized monomer characterized by its unique structure, which includes a 3,4-dihydroxyphenethyl group. This compound is designed for use in polymer synthesis, offering significant potential in the creation of functional polymers with diverse applications. Its chemical structure allows for easy incorporation into various polymer matrices, enhancing the properties of the resulting materials. The presence of hydroxyl groups contributes to increased hydrophilicity and the potential for further chemical modifications.

Application

N-(3,4-Dihydroxyphenethyl)methacrylamide is widely utilized in several key areas: Dopamine methacrylate-based materials can have a wide range of applications in biomedicine, such as drug delivery, tissue engineering, and biosensing. For example, dopamine methacrylate hydrogels have been used as a scaffold for the regeneration of injured tissues, while dopamine methacrylate coatings have been used to improve the biocompatibility and adhesion of medical implants. ; N-(3,4-Dihydroxyphenethyl)methacrylamide is valuable for creating polymer adhesives and surface coating polymers containing a catechol group. For instance, it is utilized in the development of wound dressing materials as well as in dry and wet adhesives ;

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service